missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartobind™ Rapid A Lab

Shop All Sartorius Products


Click to view available options
Quantity:
1 Unit
4 Units
Unit Size:
1 pack
Pack of 4
Description
- High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes.
- Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity.
- Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration.
- Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system.
- Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production.
Specifications
Specifications
| Flow Rate | 2.5 to 25 mL/min. |
| Column Type | Affinity |
| Stationary Phase | Agarose or Protein A |
| Diameter (Metric) Inner Column | 25 mm |
| Volume (Metric) | 0.5 mL |
| Quantity | 4 Units |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction