Learn More
Arcis Sample Prep Kit
Arcis Sample Prep Kit facilitates rapid DNA and RNA extraction in under 3 minutes
Brand: Techne 13204-52
This item is not returnable.
View return policy
Description
- Rapid DNA/RNA extraction (less than 3 minutes)
- Suitable for 50 reactions
- No instrumentation required
- Simple, stable process without heating, cooling, spin, or wash steps
- Two-step, two-tube protocol (lysis and buffer)
- DNA stable for 2 weeks
- Extract is suitable for use in PCR and qPCR reactions using standard and fast cycling protocols
- Shipped and stored at ambient temperature
Specifications
DNA/RNA Extraction Kit | |
Two-tube System (Lysis and Buffer) | |
Lysis and buffer |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Arcis Sample Prep Kit > Capacity: 50 Reactions
Spot an opportunity for improvement?Share a Content Correction