Learn More
Trajan Scientific and Medical™ XCHANGE™ Syringes for use with MEPS BINS
With PTFE-tipped plunger
Brand:  Trajan Scientific and Medical™ 2928600
 View more versions of this product
Product Code. 15219273
  • 135.00 € / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • For use on CTC PAL-xt
Accurate and reproducible injectionsXCHANGE Syringes specifically designed for MEPS Applications
Specifications
Specifications
PTFE (Plunger)
6.7 mm syringe barrel
250 μL