missing translation for 'onlineSavingsMsg'
Learn More

Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11324707
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorinating agent

Used as Fluorinating agent and oxidizing agent, gaseous etchant for silicon, particularly in the production of microelectromechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-36-9
Molecular Formula F2Xe
Molecular Weight (g/mol) 169.29
MDL Number MFCD00040538
InChI Key IGELFKKMDLGCJO-UHFFFAOYSA-N
Synonym xenon difluoride, xenon fluoride, xenon fluoride xef2, unii-6poj14981p, bis fluoranyl xenon, xef2, acmc-20aj9j, xenon difluoride trace metals basis
PubChem CID 83674
IUPAC Name difluoroxenon
SMILES F[Xe]F

Specifications

Melting Point 127°C to 129°C
Color White
Physical Form Crystalline Granules
Quantity 2 g
UN Number UN3087
Merck Index 14,10072
Solubility Information Decomposes in water.
Formula Weight 169.3
Percent Purity 99.5%
Sensitivity Moisture sensitive
Density 4.32
Assay (metals basis)
Chemical Name or Material Xenon difluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT31J023 Certificate TypeCertificate of Analysis Date09/08/2023 Catalog Number039739.09,039739.18,039739.04
Lot NumberQ21J009 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog Number039739.09,039739.18,039739.04
Lot NumberF17P23 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number039739.09,039739.18,039739.04
Lot NumberC23Q19 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number039739.09,039739.18,039739.04
Lot NumberY13F017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039739.09,039739.18,039739.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.