missing translation for 'onlineSavingsMsg'
Learn More

Cytiva XK Chromatography Columns (Empty)

Ensure trouble-free operation for low to medium pressure. Cytiva XK Chromatography Columns (Empty) connect directly to ÄKTA design instruments or other liquid chromatography systems. The user-friendly ensures reproducible, accurate packing.

Brand:  Cytiva 28-9889-50

885.40 EUR valid until 2025-12-31
Use promo code "24353" to get your promotional price.


 View more versions of this product

Product Code. 11390332

  • 932.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

XK columns are constructed of robust borosilicate glass, which exhibits excellent chemical resistance and is compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules. The clear tubes allow for visual inspection of the media bed and are available in a broad range of column dimensions for use with many different purification needs.

Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations.

  • Plunger design to ensure a homogeneous buffer distribution and reproducible packing
  • Adapter QuickLock mechanism that facilitates column handling and cleaning
  • Fittings for direct connection to chromatographiy systems
  • Cooling jacket for sensitive purifications
  • Extensive range of spare parts available, facilitating maintenance
TRUSTED_SUSTAINABILITY
Specifications

Specifications

XK Chromatography Columns (Empty)
26 mm
Inlet/Outlet (i.d.): 0.75mm
5 bar
Polypropylene (reinforced glass fiber)/Polyamide Net
Liquid chromatography at low to medium pressure
Bed height (one adapter): 530 to 650mm
Bed height (two adapters): 385 to 650mm
700 mm
Borosilicate glass
10μm pore size
Empty Chromatography Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol.

Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids.