Learn More
Tocris Bioscience™ XL 184
Potent VEGFR inhibitor; also inhibits other RTKs
Brand:  Tocris Bioscience™ 5422/50
 View more versions of this product
Product Code. 18734909
  • 1015.00 € / 50mg
Explore more special offers
Description
Description
XL 184 is a potent VEGFR inhibitor (IC50 = 0.035 nM); also inhibits c-Met, KIT, RET, FLT4, AXL, FLT3, FLT1 and Tie2 (IC50 values are 1.3, 4.6, 5.2, 6, 7, 11.3, 12 and 14.3 nM, respectively). Induces intratumoral hypoxia and apoptosis. Reduces tumor invasion and metastasis in vivo. Antiangiogenic.
Specifications
Specifications
>98%
50 mg
501.51
Cabozantinib, BMS 907351
XL 184
XL 184