Learn More
Thermo Scientific™ XLD Agar (Dehydrated)
Isolate Salmonella and Shigella from food and clinical specimens with this selective medium.
Brand: Thermo Scientific™ CM0469R
Description
Isolate Salmonella and Shigella from clinical specimens and foods with selective Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated). The medium relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of shigellae and salmonellae from non-pathogenic bacteria. The careful addition of sodium desoxycholate inhibits the growth of coliforms without decreasing the ability to support shigellae and salmonellae.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
Use XLD Medium for the screening of samples containing mixed flora suspected of harboring enteric pathogens, e.g., medical specimens or food products.
- Easy-to-read: easily differentiate Salmonella (red colonies with black centers) from Shigella (red colonies) and other coliforms (yellow, opaque colonies).
- Superior sensitivity and selectivity: exceeds that of the traditional plating media e.g., Eosin Methylene Blue and Bismuth Sulphite Agars, which tend to suppress the growth of shigellae.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
XLD Medium | |
Dehydrated Culture Media | |
For 47.0L medium |
Powder | |
2.5 kg |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6236021 | Certificate of Analysis | 12/03/2025 | CM0469R |
6205651 | Certificate of Analysis | 13/12/2024 | CM0469R |
6195045 | Certificate of Analysis | 21/11/2024 | CM0469R |
6180192 | Certificate of Analysis | 20/11/2024 | CM0469R |
6171699 | Certificate of Analysis | 23/09/2024 | CM0469R |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only