missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ xMAP™ Reporter Control Microspheres (CON2)
xMAP™ Reporter Control Microspheres (CON2), are polystyrene microspheres that are internally labeled with fluorescent dyes.
Brand: Invitrogen™ L100CON2
This item is not returnable.
View return policy
Description
xMAP™ Reporter Control Microspheres (CON2), are polystyrene microspheres that are internally labeled with fluorescent dyes. These Control Microspheres are intended to verify the calibration and optical integrity for the Luminex 200™ System. Reporter Control Microspheres verify the reporter channel.
The Luminex™ 200™ System is sold separately.
Specifications
Reporter Channel (RP1) | |
Store at 2°C to 8°C. | |
Calibration Microspheres | |
Wet Ice |
Luminex™ Instruments, Luminex™ 200™ System | |
Luminex™ | |
5 mL |
For Research Use Only. Not for use in diagnostic procedures.