Learn More
Grant Instruments™ XUB Series Digital Ultrasonic Baths
The XUB range of digitally controlled benchtop ultrasonic baths offer consistent and reliable performance in a variety of environments. Incorporating Frequency LEAP technology to ensure uniform levels of ultrasonic activity throughout the fluid, these baths offer high performance giving an accurate and precise ultrasonic process.
2350.00 € - 3645.00 €
Specifications
Timer | 0 to 99 min. |
---|---|
Voltage | 230 V |
Temperature (Metric) Range | Ambient +5°C to 70°C |
Operating Frequency | 32 to 38 kHz |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11470998
|
Grant Instruments™
XUB18 |
17.5 L |
3120.00 €
Each |
In Stock
Log in to see stock. |
|||||
11480998
|
Grant Instruments™
XUB25 |
25 L |
3645.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
11460998
|
Grant Instruments™
XUB12 |
12.5 L |
2790.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
11440998
|
Grant Instruments™
XUB5 |
4.5 L |
2350.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
The XUB range of digitally controlled benchtop ultrasonic baths offer consistent and reliable performance in a variety of environments. Incorporating Frequency LEAP technology to ensure uniform levels of ultrasonic activity throughout the fluid, these baths offer high performance giving an accurate and precise ultrasonic process.
- Adjustable speed control from 500 to 3000 rpm
- Handles up to 32 tubes in three different sizes/combinations
- Additional VP-1 flat head for single tube mixing supplied with unit
- Rubber suction pads hold tight to the work surface and prevent the unit from 'walking' - they also absorb vibration and prevent its transmission to the workbench
- Easy to operate - select either 'Continuous' or 'Touch' operation and turn the knob to adjust the speed
- The 32-socket universal platform for three tubes sizes (16 x 1.5mL, 8 x 0.5mL, 8 x 0.2mL) and a tip for mixing individual tubes up to 15mL
- Operating temperature range: + 4°C to 40°C
Specifications
0 to 99 min. | |
230 V | |
Ambient +5°C to 70°C | |
32 to 38 kHz |
Safety and Handling
- Grant M2 ultrasonic solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : C9XYLATE, QUATERNARY FATTY AMINE ETHOXYLATE, BENZENESULFONIC ACID
Your input is important to us. Please complete this form to provide feedback related to the content on this product.