missing translation for 'onlineSavingsMsg'
Learn More

Xylene, BAKER ANALYZED, J.T. Baker™

Product Code. 10587082
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

pH Liquid
Melting Point -34°C
CAS 1330-20-7
Color Colorless
Density 0.87g/cm3 at 20°C
Boiling Point 137°C to 143°C
Quantity 2.5 L
Explosion Limit 1.1 - 7 Vol %
Flash Point 24°C
Ignition Point 500°C
Odor Aromatic
Packaging Glass
Residue after Evaporation Max. 0.002%
Linear Formula C6H4(CH3)2
Viscosity 0.61 mPas at 20°C
Water Max. 0.03%
Synonym Xylol
Solubility Information 0.2g/L in water
Percent Purity ≥98.5%
Grade Exceeds ACS specifications
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.