missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, extra pure, mixture of isomers, Thermo Scientific Chemicals

53.67 € - 115.00 €

Chemical Identifiers

CAS 1330-20-7, 100-41-4
Molecular Formula C8H10
Synonym Dimethylbenzene
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10427413
SDS View Documents
Thermo Scientific Acros
390790010
1 L Glass Bottle
53.67 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10479233
SDS View Documents
Thermo Scientific Acros
390790025
2.5 L Glass Bottle
93.26 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11904451
SDS View Documents
Thermo Scientific Acros
390790100
10 L Plastic Drum N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xylenes, extra pure, mixture of isomers, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.