missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, puriss. p.a., ACS reagent, Honeywell
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
25 L
144 x 1 L
Specifications
Specifications
Density | 0.86g/mL at 25°C |
Quantity | 25 L |
Explosion Limit | 0.07 |
Flash Point | 25°C (77°F) |
Linear Formula | C6H4(CH3)2 |
Vapor Pressure | 18 mmHg (37.7°C) |
UN Number | UN1307 |
Formula Weight | 106.17g/mol |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Xylenes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'