missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Yeast Extract
Click to view available options
Quantity:
100 g
500 g
Descripción
Yeast extract is used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Form | Powder |
Packaging Type | Glass Bottle |
Quantity | 100 g |
Sensitivity | Hygroscopic |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido