Learn More
Thermo Scientific Chemicals Yeast Extract
Used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.
101.51 € - 175.88 €
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15483079
|
Thermo Scientific Chemicals
J60287.36 |
500 g |
101.51 €
500g |
In Stock
Log in to see stock. |
||||||
15493079
|
Thermo Scientific Chemicals
J60287.A1 |
1 kg |
175.88 €
1kg |
In Stock
Log in to see stock. |
||||||
Description
Microbial mediaYeast extract is used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Powder | |
Plastic Bottle |
Safety and Handling
missing translation for 'einecsNumber' : 232-387-9
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.