missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast Hydrolysate Enzymatic, MP Biomedicals™

an autolyzed yeast extract that is a concentrated source of soluble protein components, carbohydrates and B-complex vitamins that are natural to yeast.

Brand:  MP Biomedicals™ 0210330480

Product Code. 11450655

  • 2655.00 € / 100lb
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Produced from 100% brewer’s yeast.
  • Brown-colored powder, soluble in water
  • Store at room temperature
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brown
Store at Room Temperature
Source of soluble protein components; carbohydrates; and B Complex vitamins
Yeast hydrolysate enzymatic
Powder
100 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.