Learn More
Checkers Yellow Jacket™ 3-Channel AMS Cable Protector Kit
Yellow Jacket™ 3-channel AMS cable protector kit features a gradual slope and slip-resistant surface to ensure a safer crossing point for carts, forklifts, vehicles with small wheels and heavy trucks.
Brand:  Checkers PYJS0003YB
Product Code. 17762428
  • 2860.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Yellow Jacket™ 3-channel AMS cable protector kit features a gradual slope and slip-resistant surface to ensure a safer crossing point for carts, forklifts, vehicles with small wheels and heavy trucks
  • In addition, each Accessibility Ramp system is manufactured to meet ADA and DDA compliant specifications
Specifications
Specifications
6.5 ft.
181 cm
Polyethylene
198 cm
Black, Yellow
Cable Protector Kit