missing translation for 'onlineSavingsMsg'
Learn More
Learn More
This item is not returnable.
View return policy
Specifications
10 L | |
Bath Filler | |
10 L |
30°C to 200°C | |
Ultra 350 | |
47mm2/sec. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bath liquid ULTRA 350 (1 can à 10 litres) .
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dibenzylbenzene, ar-methyl derivitive