Learn More
YSI™ Ammonium Calibration Solutions
Extremely accurate calibration solutions to calibrate field instruments equipped with ammonium ISE sensors
Brand:  YSI™ 003841
 View more versions of this product
Product Code. 12786945
  • 91.75 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Concentration: 1mg/mL
  • Packaging: Plastic bottle
Order Info
Dated Material. Item is heat packed in the winter months.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Concentration: 1mg/L
Ammonium Calibration Solution
500 mL
Calibrating field ammonium ISE sensors