missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Confidence Solution, YSI™
Click to view available options
Water monitoring solution; 475mL
Description
- This solution is designed for verifying measurements, not calibration
- Simply immerse multiparameter probes in the solution and verify the pH, ORP, and conductivity values instantly
Specifications
Specifications
Packaging | 475mL |
Quantity | 475 mL |
Analyte or Component Names | Confidence Solution |
Chemical Name or Material | Confidence Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'