missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Zobell ORP Calibration Solution

For calibrating ORP/Redox on several YSI instruments

Brand:  YSI™ 061320

Product Code. 12744526

  • 40.15 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • A convenient temperature chart with corresponding ORP values is on the side of the container for simple calibration value inputs
  • Concentrated power makes 125mL of calibration standard
  • Plastic bottle (125mL)

Expiration Dates:
Unmixed: 2 years
Mixed: 6 months

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calibration Solution
ORP calibration solution
119 mL
Calibrating ORP/Redox on YSI™ instruments
Temperature chart with corresponding ORP values is on the side of the container
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.