missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium foil, 0.3mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11325928
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈1.3g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Ytterbium foil, 0.3mm (0.01 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'