missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11345128
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226944-34-9
Molecular Formula YbBr3·xH2O
MDL Number MFCD00049613
Synonym ytterbium bromide hydrate, acmc-20aj9h, ytterbium 3+ bromide-water 1/3/1

Specifications

Physical Form Lump
Quantity 10 g
Formula Weight 412.77 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Ytterbium (III) bromide hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'