missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10607504
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-01-5
Molecular Formula Cl3Yb·6H2O
Molecular Weight (g/mol) 387.48
MDL Number MFCD00149885

Specifications

Melting Point 865.0°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.90%
Linear Formula YbCl3·6H2O
Fieser 16,384
Merck Index 14, 10105
Solubility Information Solubility in water: soluble. Other solubilities: soluble in absolute alcohol
Formula Weight 387.48
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) chloride hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ytterbium(III) chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.