missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
50 g
Description
Ytterbium(III) fluoride is used in the preparation and study of fluorinated glasses. Further, it is employed as a laboratory reagent and in optical glasses and structural ceramics. It finds application in catalysts, electrical components and photo-optical materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1157°C |
Physical Form | Lump/Powder |
Quantity | 10 g |
UN Number | UN3288 |
Solubility Information | Slightly soluble in strong mineral acids. Insoluble in water. |
IUPAC Name | ytterbium(3+);trifluoride |
Formula Weight | 230.04 |
Percent Purity | 99.99% |
Grade | Reacton™ |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'