missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals

83.31 € - 320.60 €

Chemical Identifiers

CAS 1314-37-0
Molecular Formula O3Yb2
Molecular Weight (g/mol) 394.09
MDL Number MFCD00011290
InChI Key UZLYXNNZYFBAQO-UHFFFAOYSA-N
IUPAC Name diytterbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Yb+3].[Yb+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351448
View Documents
Thermo Scientific Alfa Aesar
010919.06
5 g
83.31 €
5g
Please to purchase this item. Need a web account? Register with us today!
11341448
View Documents
Thermo Scientific Alfa Aesar
010919.14
25 g
320.60 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It serves as a dopant for silicate mineral crystals in lasers, and a colorant for glasses and enamels. It is employed in the preparation of optical fibers, and special types of alloys. It is used in the preparation of chiral ytterbium complexes which are used as catalysts for enantioselective organic synthesis with high yields. It finds application in optic, electronic and ceramic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-37-0
394.09
UZLYXNNZYFBAQO-UHFFFAOYSA-N
[O--].[O--].[O--].[Yb+3].[Yb+3]
O3Yb2
MFCD00011290
diytterbium(3+) trioxidandiide

Specifications

2227°C
Powder
O3Yb2
Soluble in hot dilute acids. Insoluble in water.
[O--].[O--].[O--].[Yb+3].[Yb+3]
394.09
9.2
1314-37-0
99.998% (REO)
MFCD00011290
UZLYXNNZYFBAQO-UHFFFAOYSA-N
diytterbium(3+) trioxidandiide
394.08
Ytterbium(III) oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-234-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.