missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Product Code. 11420311
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13498-08-3
Molecular Formula Cl3H2O13Yb
Molecular Weight (g/mol) 489.41
MDL Number MFCD00049612
InChI Key TYYKUTNLJJSRBB-UHFFFAOYSA-K
Synonym ytterbium iii perchlorate, acmc-1bqma, yb clo4 3, trihyperchloric acid ytterbium salt, ytterbium iii perchlorate solution, perchloric acid,ytterbium 3+ salt 8ci,9ci
PubChem CID 11363421
SMILES O.[Yb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Insoluble in water.
IUPAC Name ytterbium(3+) hydrate triperchlorate
Formula Weight 579.48 (471.39 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Ytterbium(III) perchlorate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'