Learn More
Ytterbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™
63.00 € - 274.00 €
Chemical Identifiers
CAS | 13498-08-3 |
---|---|
Molecular Formula | Cl3H2O13Yb |
Molecular Weight (g/mol) | 489.41 |
MDL Number | MFCD00049612 |
InChI Key | TYYKUTNLJJSRBB-UHFFFAOYSA-K |
Synonym | ytterbium iii perchlorate, acmc-1bqma, yb clo4 3, trihyperchloric acid ytterbium salt, ytterbium iii perchlorate solution, perchloric acid,ytterbium 3+ salt 8ci,9ci |
PubChem CID | 11363421 |
SMILES | O.[Yb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13498-08-3 | |
489.41 | |
TYYKUTNLJJSRBB-UHFFFAOYSA-K | |
11363421 |
Cl3H2O13Yb | |
MFCD00049612 | |
ytterbium iii perchlorate, acmc-1bqma, yb clo4 3, trihyperchloric acid ytterbium salt, ytterbium iii perchlorate solution, perchloric acid,ytterbium 3+ salt 8ci,9ci | |
O.[Yb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Specifications
13498-08-3 | |
Cl3H2O13Yb | |
UN3211 | |
TYYKUTNLJJSRBB-UHFFFAOYSA-K | |
ytterbium(3+);triperchlorate | |
11363421 | |
Reagent | |
Odorless |
Liquid | |
MFCD00049612 | |
ytterbium iii perchlorate, acmc-1bqma, yb clo4 3, trihyperchloric acid ytterbium salt, ytterbium iii perchlorate solution, perchloric acid,ytterbium 3+ salt 8ci,9ci | |
O.[Yb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
489.41 | |
579.48 (471.39 Anhydrous) | |
50% w/w aq. soln. | |
Ytterbium(III) perchlorate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.