missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) sulfate octahydrate, REacton™, 99.9% (REO)

Product Code. p-7049511
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11362518

Brand: Thermo Scientific Alfa Aesar 014572.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10034-98-7 | H16O20S3Yb2 | 778.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-98-7
Molecular Formula H16O20S3Yb2
Molecular Weight (g/mol) 778.40
MDL Number MFCD00149887
InChI Key MDOSIOHMOWGVIM-UHFFFAOYSA-H
Synonym unii-e5w6do92vs, e5w6do92vs, 3so4.2yb.8h2o, ytterbium sulfate,, ytterbium sulfate octahydrate, ytterbium sulfate, ytterbium iii sulfate octahydrate, reacton, diytterbium 3+ octahydrate trisulfate, sulfuric acid,ytterbium 3+ salt 3:2 , octahydrate 8ci,9ci
PubChem CID 25022290
SMILES O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 10 g
Merck Index 14,10105
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
IUPAC Name ytterbium(3+);trisulfate;octahydrate
Formula Weight 778.38 (634.26 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Ytterbium(III) sulfate octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.