missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals Product Code.: 11663009

Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Product Code. 11663009
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11663009

Brand: Thermo Scientific Acros 434050050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54761-04-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.25
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate
PubChem CID 2733225
IUPAC Name trifluoromethanesulfonate;ytterbium(3+)
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3]

Specifications

Quantity 5 g
Assay Percent Range 26.8 to 29% (Yb) (Complexometry)
Linear Formula (CF3SO3)3Yb
Merck Index 15,10304
Solubility Information Solubility in water: soluble
Formula Weight 620.25
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) triflate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467691 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number434050050,434050010
Lot NumberA0462293 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number434050050,434050010
Lot NumberA0443473 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number434050050,434050010
Lot NumberA0398594 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number434050050,434050010
Lot NumberA0400313 Certificate TypeCertificate of Analysis Date30/08/2021 Catalog Number
compliance-icons
Product Identifier
  • Ytterbium(III) triflate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.