missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

23.22 € - 218.09 €

Chemical Identifiers

CAS 252976-51-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.23
MDL Number MFCD06200261
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate
PubChem CID 22836430
IUPAC Name ytterbium(3+) tritrifluoromethanesulfonate
SMILES [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10646871
View Documents
Thermo Scientific Acros
304170010
1 g Glass bottle
23.22 €
1g
Please to purchase this item. Need a web account? Register with us today!
10085863
View Documents
Thermo Scientific Acros
304170050
5 g Glass bottle
59.19 €
5g
Please to purchase this item. Need a web account? Register with us today!
10045913
View Documents
Thermo Scientific Acros
304170250
25 g Glass bottle
218.09 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

252976-51-5
620.23
AHZJKOKFZJYCLG-UHFFFAOYSA-K
22836430
[Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
C3F9O9S3Yb
MFCD06200261
ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate
ytterbium(3+) tritrifluoromethanesulfonate

Specifications

118.0°C to 122.0°C
White
99.90%
(CF3SO3)3Yb·xH2O
ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate
AHZJKOKFZJYCLG-UHFFFAOYSA-K
ytterbium(3+) tritrifluoromethanesulfonate
22836430
99.9%
Ytterbium(III) triflate hydrate
252976-51-5
Crystalline Powder
C3F9O9S3Yb
MFCD06200261
Solubility in water: soluble
[Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
620.23
620.23
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.