missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium pieces, sublimed, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15491897
Click to view available options
Quantity:
5 g
25 g
100 g

May contain up to 0.15% Calcium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Color Silver to Gray
Quantity 5 g
Assay Percent Range 99.9%
Formula Weight 173.04
Odor Odorless
Appearance Silver Gray pieces
Chemical Name or Material Ytterbium, May contain up to 0.15% Calcium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'