Learn More
Ytterbium pieces, sublimed, 99.9% (REO), Thermo Scientific Chemicals
May contain up to 0.15% Calcium
99.50 € - 1024.00 €
Chemical Identifiers
CAS | 7440-64-4 |
---|---|
Molecular Formula | Yb |
Molecular Weight (g/mol) | 173.05 |
MDL Number | MFCD00011286 |
InChI Key | NAWDYIZEMPQZHO-UHFFFAOYSA-N |
Synonym | unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n |
PubChem CID | 23992 |
ChEBI | CHEBI:33381 |
IUPAC Name | ytterbium |
SMILES | [Yb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15491897
|
Thermo Scientific Alfa Aesar
043431.06 |
5 g |
99.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15401907
|
Thermo Scientific Alfa Aesar
043431.14 |
25 g |
325.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15411907
|
Thermo Scientific Alfa Aesar
043431.22 |
100 g |
1024.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-64-4 | |
173.05 | |
NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
23992 | |
ytterbium |
Yb | |
MFCD00011286 | |
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
CHEBI:33381 | |
[Yb] |
Specifications
7440-64-4 | |
99.9% | |
MFCD00011286 | |
NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
ytterbium | |
23992 | |
173.04 | |
Silver Gray pieces |
Silver to Gray | |
Yb | |
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
[Yb] | |
173.05 | |
CHEBI:33381 | |
Odorless | |
Ytterbium, May contain up to 0.15% Calcium |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II
missing translation for 'einecsNumber' : 231-173-2
missing translation for 'rtecsNumber' : ZG1925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only