Learn More
Thermo Scientific Chemicals Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Brand:  Thermo Scientific Chemicals 041274.KF
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15463825
  • 456.27 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zirconium Oxide
10 mL
43mm
1 Piece
24mm
Yttria Stabilized Zirconium Oxide Cylindrical Crucibles