missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Specifications
Color | White |
Physical Form | Crystalline Powder and Lumps |
Quantity | 25 g |
Assay Percent Range | 99.90% |
Solubility Information | Solubility in water: soluble |
IUPAC Name | yttrium(3+) triacetate |
Formula Weight | 266.04 |
Percent Purity | 99.9% |
Grade | Trace Metal Basis |
Infrared Spectrum | Authentic |
Show More |
Safety and Handling
Product Identifier
- Yttrium acetate hydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Yttrium acetate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction