missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium Aluminum lump, 99.9% (metals basis excluding Ca), Ca 1% max, Thermo Scientific Chemicals
128.64 € - 375.87 €
Specifications
Chemical Name or Material | Yttrium Aluminum lump |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Y:Al; 87:12 wt% |
Assay Percent Notes | (metals basis excluding Ca), Ca 1% max. |
MDL Number | MFCD01074900 |
Percent Purity | 99.9% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302038
|
Thermo Scientific Alfa Aesar
012949.14 |
25 g |
128.64 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312038
|
Thermo Scientific Alfa Aesar
012949.22 |
100 g |
375.87 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yttrium Aluminum lump | |
(metals basis excluding Ca), Ca 1% max. | |
99.9% |
Y:Al; 87:12 wt% | |
MFCD01074900 | |
Lump |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title