missing translation for 'onlineSavingsMsg'
Learn More

Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15461707
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Rare earth element doped yttrium aluminum oxide is used in solid state lasers

Rare earth element doped yttrium aluminum oxide is used in solid state lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-21-9
MDL Number MFCD00213958
Synonym Yttrium aluminum garnet; YAG

Specifications

Melting Point 1760°C
Physical Form Powder
Quantity 25 g
Formula Weight 593.62
Percent Purity 99.9%
Refractive Index 1.833
Density 4.65 g/mL
Assay (REO)
Chemical Name or Material Yttrium aluminum oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'