missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Cobalt lump, REacton™, 99.9% (REO), Thermo Scientific Chemicals

126.63 € - 422.10 €

Specifications

Chemical Name or Material Yttrium Cobalt lump
Avg. Mol. Wt. or Mol. Wt. Range Y:Co; 84:15 wt% (Ca 1%)
Assay Percent Notes (REO)
MDL Number MFCD00209676
Percent Purity 99.9%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352038
View Documents
Thermo Scientific Alfa Aesar
012952.14
25 g
126.63 €
25g
Please to purchase this item. Need a web account? Register with us today!
11342038
View Documents
Thermo Scientific Alfa Aesar
012952.22
100 g
422.10 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Yttrium Cobalt lump
(REO)
99.9%
Lump
Y:Co; 84:15 wt% (Ca 1%)
MFCD00209676
Reacton™
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only