Learn More
Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals
169.00 € - 474.00 €
Chemical Identifiers
CAS | 7440-65-5 |
---|---|
Molecular Formula | Y |
Molecular Weight (g/mol) | 88.91 |
MDL Number | MFCD00011468 |
InChI Key | VWQVUPCCIRVNHF-UHFFFAOYSA-N |
Synonym | yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element |
PubChem CID | 23993 |
ChEBI | CHEBI:33331 |
IUPAC Name | yttrium |
SMILES | [Y] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381799
|
Thermo Scientific Alfa Aesar
012443.FF |
25 x 25 mm |
169.00 €
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400290
|
Thermo Scientific Alfa Aesar
012443.FI |
50 x 50 mm |
474.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-65-5 | |
88.91 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
23993 | |
yttrium |
Y | |
MFCD00011468 | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
CHEBI:33331 | |
[Y] |
Specifications
7440-65-5 | |
MFCD00011468 | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
[Y] | |
88.91 | |
CHEBI:33331 | |
Odorless | |
Foil | |
Yttrium foil, 0.025mm (0.001 in.) thick |
Y | |
UN3178 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
yttrium | |
23993 | |
99.9% | |
≈0.07g/25x25mm | |
(REO) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378s
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 231-174-8
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only