missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium foil, 0.1mm (0.004in) thick, 99.6% (REO)
missing translation for 'orderingAttributeHoverText'
Quantity:
25 x 25 mm
50 x 50 mm
missing translation for 'unitSize'
25mm
Each
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Spezifikation
Spezifikation
| Quantity | 50 x 50 mm |
| Odor | Odorless |
| Packaging | Mylar under Argon |
| Weight | ≈0.28g/25x25mm |
| Form | Foil |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Chemical Name or Material | Yttrium foil, 0.1mm (0.004 in.) thick |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur