missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) acetate tetrahydrate, 99.9% (REO)

Product Code. p-7049506
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11372508

Brand: Thermo Scientific Alfa Aesar 014565.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In analytical chemistry | CAS: 85949-60-6 | C6H17O10Y | 338.10 g/mol

In analytical chemistry.Yttrium(III) acetate tetrahydrate is used in analytical chemistry and in catalysis. It is used in the preparation of yttrium trifluoride nanoparticles, which shows antibacterial properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85949-60-6
Molecular Formula C6H17O10Y
Molecular Weight (g/mol) 338.10
MDL Number MFCD00013048
InChI Key AIQRTHPXPDTMBQ-UHFFFAOYSA-K
IUPAC Name yttrium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula Y(OOCCH3)3·4H2O
Solubility Information Soluble in hot water.
Formula Weight 338.10 (266.04 Anhydrous)
Percent Purity 99.9%
Density 1.5 g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.