missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

95.48 € - 294.47 €

Chemical Identifiers

CAS 85949-60-6
Molecular Formula C6H17O10Y
Molecular Weight (g/mol) 338.10
MDL Number MFCD00013048
InChI Key AIQRTHPXPDTMBQ-UHFFFAOYSA-K
IUPAC Name yttrium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366208
SDS View Documents
Thermo Scientific Alfa Aesar
011186.14
25 g
95.48 €
25g
Please to purchase this item. Need a web account? Register with us today!
11356208
SDS View Documents
Thermo Scientific Alfa Aesar
011186.22
100 g
294.47 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttrium(III) acetate tetrahydrate is used as laboratory reagents, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

85949-60-6
338.10
AIQRTHPXPDTMBQ-UHFFFAOYSA-K
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
C6H17O10Y
MFCD00013048
yttrium(3+) triacetate tetrahydrate

Specifications

(decomposition)
Crystalline Aggregates
C6H17O10Y
Soluble in Hot water.
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
338.10
1.5 g/mL
85949-60-6
99.99% (REO)
MFCD00013048
AIQRTHPXPDTMBQ-UHFFFAOYSA-K
yttrium(3+) triacetate tetrahydrate
338.10 (266.04 Anhydrous)
Yttrium(III) acetate tetrahydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 245-612-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.