missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11313467
Click to view available options
Quantity:
10 g
50 g
250 g

Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-92-9
Molecular Formula Cl3Y
MDL Number MFCD00011470

Specifications

Melting Point 721°C
Quantity 10 g
Assay Percent Range 99.9%
Solubility Information Soluble in water,ethanol and pyridine.
Formula Weight 195.26
Sensitivity Hygroscopic
Density 2.67
Chemical Name or Material Yttrium(III) Chloride, Rare Earth Oxides Content

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'