missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

60.70 € - 409.04 €

Chemical Identifiers

CAS 12741-05-8
Molecular Formula Cl3Y
Molecular Weight (g/mol) 195.26
MDL Number MFCD00149941
InChI Key PCMOZDDGXKIOLL-UHFFFAOYSA-K
Synonym yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
PubChem CID 57347377
IUPAC Name yttrium(3+);trichloride;hydrate
SMILES [Cl-].[Cl-].[Cl-].[Y+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316208
SDS View Documents
Thermo Scientific Alfa Aesar
011185.14
25 g
60.70 €
25g
Please to purchase this item. Need a web account? Register with us today!
11336208
SDS View Documents
Thermo Scientific Alfa Aesar
011185.22
100 g
191.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
11346208
SDS View Documents
Thermo Scientific Alfa Aesar
011185.30
250 g
409.04 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttrium(III) chloride is used a precursor for the synthesis of optically active crystals and as a starting material for the synthesis of polymerization catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12741-05-8
195.26
PCMOZDDGXKIOLL-UHFFFAOYSA-K
57347377
[Cl-].[Cl-].[Cl-].[Y+3]
Cl3Y
MFCD00149941
yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
yttrium(3+);trichloride;hydrate

Specifications

12741-05-8
Crystalline Aggregates
YCl3·xH2O
14,10107
Soluble in water,pyridine,ethanol.
[Cl-].[Cl-].[Cl-].[Y+3]
195.26
195.26 (Anhydrous)
Reacton™
Hygroscopic
(REO)
Pink to White
Cl3Y
MFCD00149941
yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
PCMOZDDGXKIOLL-UHFFFAOYSA-K
yttrium(3+);trichloride;hydrate
57347377
99.99%
Odorless
2.18g/mL
Yttrium(III) chloride hydrate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-801-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.