missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

For vacuum deposition

79.19 € - 967.82 €

Chemical Identifiers

CAS 13709-49-4
Molecular Formula F3Y
Molecular Weight (g/mol) 145.90
MDL Number MFCD00049714
InChI Key RBORBHYCVONNJH-UHFFFAOYSA-K
IUPAC Name yttrium(3+) trifluoride
SMILES [F-].[F-].[F-].[Y+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354128
SDS View Documents
Thermo Scientific Alfa Aesar
041424.09
10 g
79.19 €
10g
Please to purchase this item. Need a web account? Register with us today!
11364128
SDS View Documents
Thermo Scientific Alfa Aesar
041424.18
50 g
265.32 €
50g
Please to purchase this item. Need a web account? Register with us today!
11374128
SDS View Documents
Thermo Scientific Alfa Aesar
041424.30
250 g
967.82 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttrium(III) fluoride is used for the production of metallic yttrium, thin films, glasses, electronics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-257-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.