missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 13709-49-4
Molecular Formula F3Y
Molecular Weight (g/mol) 145.90
MDL Number MFCD00049714
InChI Key RBORBHYCVONNJH-UHFFFAOYSA-K
IUPAC Name yttrium(3+) trifluoride
SMILES [F-].[F-].[F-].[Y+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366898
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013650.14
25 g N/A
N/A
11346898
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013650.22
100 g N/A
N/A
11356898
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013650.36
500 g N/A
N/A
Description

Description

Yttrium(III) fluoride is used in the production of yttrium metal, glasses and ceramics. It is also used in the preparation of low-index film layers, which exhibit good transparency in the UV through infrared regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13709-49-4
145.90
RBORBHYCVONNJH-UHFFFAOYSA-K
[F-].[F-].[F-].[Y+3]
F3Y
MFCD00049714
yttrium(3+) trifluoride

Specifications

1387°C
Lump/Powder
MFCD00049714
Soluble in acids. Insoluble in water.
[F-].[F-].[F-].[Y+3]
145.90
99.9%
Hygroscopic
Yttrium(III) fluoride, Anhydrous
13709-49-4
F3Y
UN3288
RBORBHYCVONNJH-UHFFFAOYSA-K
yttrium(3+) trifluoride
145.9
Reacton™
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-257-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only