missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) hexafluoro-2,4-pentanedionate, Thermo Scientific Chemicals

Product Code. 11347028
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18911-76-7
Molecular Formula C15H3F18O6Y
Molecular Weight (g/mol) 710.06
MDL Number MFCD00064764
InChI Key AJVDSGQQIVVYRX-UHFFFAOYSA-N
Synonym yttrium hexafluoroacetylacetonate, z-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one; yttrium
PubChem CID 50930692
SMILES [Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F

Specifications

Melting Point 166°C to 170°C
Physical Form Crystalline
Quantity 1 g
IUPAC Name yttrium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Formula Weight 710.1
Chemical Name or Material Yttrium(III) hexafluoro-2,4-pentanedionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'