Learn More
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
Description
It is used as a reagent, in optics, ceramics, glass and electronics. It is used in superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.9% (REO) |
Solubility Information | Soluble in water. |
Formula Weight | 383.01 (274.92 Anhydrous) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Yttrium(III) nitrate hexahydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.