Learn More
Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
40.90 € - 1591.00 €
Chemical Identifiers
CAS | 140191-40-8 |
---|---|
Molecular Formula | N3O9Y |
Molecular Weight (g/mol) | 274.92 |
MDL Number | MFCD00011469 |
InChI Key | BXJPTTGFESFXJU-UHFFFAOYSA-N |
Synonym | yttrium iii nitrate hydrate, yttrium nitrate hydrate, 3no3.y.h2o, nitric acid, yttrium iii salt, nitric acid, yttrium 3++ salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, reacton |
PubChem CID | 23169389 |
SMILES | [Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376208
|
Thermo Scientific Alfa Aesar
011187.06 |
5 g |
40.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306218
|
Thermo Scientific Alfa Aesar
011187.14 |
25 g |
150.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396208
|
Thermo Scientific Alfa Aesar
011187.22 |
100 g |
439.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386208
|
Thermo Scientific Alfa Aesar
011187.59 |
5 x 100 g |
1591.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It acts as a precursor for the preparation of yttrium oxide nano particles and for nanoscale coatings of carbon composites. It is an effective yttrium source for the preparation of yttrium-based surfactant mesophases which are used as adsorbing agents and in optically functional materials. It is used in optics, ceramics, glass, electronics, and superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products. Silica-supported yttrium nitrate hydrate acts as a heterogeneous catalyst for the preparation of 2-Amino-4H-Chromenes in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
140191-40-8 | |
274.92 | |
BXJPTTGFESFXJU-UHFFFAOYSA-N | |
23169389 |
N3O9Y | |
MFCD00011469 | |
yttrium iii nitrate hydrate, yttrium nitrate hydrate, 3no3.y.h2o, nitric acid, yttrium iii salt, nitric acid, yttrium 3++ salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, reacton | |
[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
140191-40-8 | |
N3O9Y | |
UN1477 | |
yttrium iii nitrate hydrate, yttrium nitrate hydrate, 3no3.y.h2o, nitric acid, yttrium iii salt, nitric acid, yttrium 3++ salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, reacton | |
BXJPTTGFESFXJU-UHFFFAOYSA-N | |
yttrium(3+);trinitrate;hydrate | |
23169389 | |
99.99% | |
Hygroscopic | |
Yttrium(III) nitrate hydrate |
Crystalline Aggregates | |
MFCD00011469 | |
14,10107 | |
Soluble in water. | |
[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
274.92 | |
274.92 (Anhydrous) | |
Reacton™ | |
(REO) |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 233-802-6
missing translation for 'rtecsNumber' : ZG3675000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only