missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals

126.63 € - 528.63 €

Chemical Identifiers

CAS 13266-82-5
Molecular Formula C6H18O21Y2
Molecular Weight (g/mol) 604.001
MDL Number MFCD00150502
InChI Key CTXJKXXQOORSKU-UHFFFAOYSA-H
PubChem CID 131872179
IUPAC Name oxalate;yttrium(3+);nonahydrate
SMILES C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.O.O.O.[Y+3].[Y+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352508
SDS View Documents
Thermo Scientific Alfa Aesar
014562.18
50 g
126.63 €
50g
Please to purchase this item. Need a web account? Register with us today!
11342508
SDS View Documents
Thermo Scientific Alfa Aesar
014562.30
250 g
528.63 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttrium oxalate is applied in ceramics, glass, and electronics. It is an important material for tri-bands rare Earth phosphors and yttrium-iron-garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.