Learn More
Yttrium(III) oxide, nanopowder, 99.995% (REO), Thermo Scientific Chemicals
99.19 € - 1144.70 €
Chemical Identifiers
CAS | 1314-36-9 |
---|---|
Molecular Formula | O3Y2 |
Molecular Weight (g/mol) | 225.81 |
MDL Number | MFCD00011473 |
InChI Key | RUDFQVOCFDJEEF-UHFFFAOYSA-N |
IUPAC Name | diyttrium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Y+3].[Y+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442877
|
Thermo Scientific Alfa Aesar
047154.14 |
25 g |
99.19 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452877
|
Thermo Scientific Alfa Aesar
047154.22 |
100 g |
297.48 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462877
|
Thermo Scientific Alfa Aesar
047154.36 |
500 g |
1144.70 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-36-9 | |
225.81 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Y+3].[Y+3] |
O3Y2 | |
MFCD00011473 | |
diyttrium(3+) trioxidandiide |
Specifications
2410°C | |
White | |
O3Y2 | |
MFCD00011473 | |
Insoluble in water. Soluble in alcohol and dilute acids | |
[O--].[O--].[O--].[Y+3].[Y+3] | |
225.81 | |
99.995% | |
5.01 g/mL | |
Yttrium(III) oxide |
1314-36-9 | |
Powder | |
Y2O3 | |
14,10107 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
diyttrium(3+) trioxidandiide | |
225.81 | |
Odorless | |
(REO) |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-233-5
missing translation for 'rtecsNumber' : ZG3850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Yttrium(III) oxide, nanopowder
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.